Learn More
Aluminum potassium sulfate dodecahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
In printing, fabrics, dyeing, manufacturing of paper, vegetable glue, baking powder, purifying water, clarifying sugar, as catalyst in ammonia synthesis, hardening agent in microscopyAluminum potassium sulfate dodecahydrate is used in leather tanning, water purification, fire proof textiles and backing powder. In the cosmetic industry, it is used as an after shave treatment, deodorant, hardener for photographic emulsions and as a mordant for dyes. In medical field, it finds application as an astringent, antiseptic and to reduce bleeding in minor cuts and abrasions and nosebleeds..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 92°C |
Color | White |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | 98 -102% |
Solubility Information | Soluble in water,glycerol. Insoluble in alcoholSoluble in water and glycerol. Insoluble in acetone and alcohol. |
Formula Weight | 474.38 (258.20 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Appearance | White granular |
Show More |
RUO – Research Use Only