missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl isocyanate, 98+%

Product Code. 11424284
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11424284

Brand: Thermo Scientific Alfa Aesar A12576.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1195-45-5 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1195-45-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00002023
InChI Key DSVGFKBFFICWLZ-UHFFFAOYSA-N
PubChem CID 70955
IUPAC Name 1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=CC=C1N=C=O)F

Specifications

Density 1.21
Boiling Point 55°C to 56°C (8 mmHg)
Flash Point 52°C (125°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.514
Quantity 5 g
UN Number UN3080
Beilstein 471609
Sensitivity Moisture sensitive; May form a precipitate
Synonym 4-fluorophenyl isocyanate, 4-fluorophenylisocyanate, p-fluorophenyl isocyanate, benzene, 1-fluoro-4-isocyanato, isocyanic acid 4-fluorophenyl ester, 1-fluoro-4-isocyanato-benzene, benzene,1-fluoro-4-isocyanato, 4-fluorobenzenisocyanate, paragos 331046, timtec-bb sbb032971
Formula Weight 137.11
Percent Purity ≥98%
Chemical Name or Material 4-Fluorophenyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Fluorophenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Fluorophenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.