missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Aluminum platform for Innova™ 44/44R
Click to view available options
For Use With (Equipment):
250 mL Erlenmeyer Flask Dedicated Platform
Unit Size:
Each
Description
Aluminum platform for Innova™ 44/44R, 76 × 46 cm, not interchangeable with other 76 × 46 cm platforms, 250 mL Erlenmeyer flask dedicated platform- Flask Capacity: 40
Specifications
Specifications
| Dimensions (L x W) | 76 x 46 cm |
| Description | Not Interchangeable With Other |
| For Use With (Equipment) | 250 mL Erlenmeyer Flask Dedicated Platform |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction