Learn More
Thermo Scientific Alfa Aesar Aluminum plate, 6.35mm (0.25 in.) thick, 99% (metals basis)
Brand: Thermo Scientific Alfa Aesar 011062.RY
Aluminum plate, 6.35mm (0.25 in.) thick, 99% (metals basis)
Code : NEW
Additional Details : CAS Number : 7429-90-5 Weight : 0.01000kg