missing translation for 'onlineSavingsMsg'
Learn More

Aluminum phosphate, pure, Thermo Scientific Chemicals

Catalog Number 10061581
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
7784-30-7
AlO4P
121.95
ILRRQNADMUWWFW-UHFFFAOYSA-K
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
64655
aluminum;phosphate
[O-]P(=O)([O-])[O-].[Al+3]
This item is not returnable. View return policy
7784-30-7
AlO4P
121.95
ILRRQNADMUWWFW-UHFFFAOYSA-K
aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
64655
aluminum;phosphate
[O-]P(=O)([O-])[O-].[Al+3]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >1500.0°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula AlPO4
Merck Index 15,352
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol, soluble in acids, alkali
Formula Weight 121.95
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Aluminum phosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'