missing translation for 'onlineSavingsMsg'
Learn More

Aluminum phosphate, Puratronic™, 99.99% (metals basis)

Product Code. p-4407201
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11300819

Brand: Thermo Scientific Alfa Aesar 010628.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7784-30-7 | AlO4P | 121.951 g/mol

Aluminum phosphate is used in the production of glass, mixture of ceramic, waterproofing concrete and dental cements, in cosmetics as a emollient, flame retardant, catalyst in organic synthesis, fireproofing textile, pharmaceuticals and dyes. It also used in many adhesives, binders and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-30-7
Molecular Formula AlO4P
Molecular Weight (g/mol) 121.951
MDL Number MFCD00003430
InChI Key ILRRQNADMUWWFW-UHFFFAOYSA-K
Synonym aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
PubChem CID 64655
IUPAC Name aluminum;phosphate
SMILES [O-]P(=O)([O-])[O-].[Al+3]

Specifications

Melting Point >1500°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in concentrated hydrochloric acid,alcohol and nitric acid. Insoluble in water.
Formula Weight 121.95
Appearance White powder
Density 2.566 g/cm3
Chemical Name or Material Aluminum phosphate
compliance-icons
Product Identifier
  • Aluminum phosphate, Puratronic
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.