missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Perchlorate Nonahydrate, 98%, Honeywell™ Fluka™

Product Code. 15631840 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15631840 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15631840 Supplier Honeywell-Fluka Supplier No. 208485100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

>98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 81029-06-3
Physical Form Solid
Quantity 100 g
Molecular Formula AlCl3H18O21
Linear Formula Al(ClO4)3·9H2O
MDL Number MFCD00149130
UN Number UN1481
Synonym aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
InChI Key CYRDFMGMNQJULY-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]
IUPAC Name aluminum;triperchlorate;nonahydrate
Molecular Weight (g/mol) 487.455
PubChem CID 16211522
Formula Weight 487.47g/mol
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Aluminum Perchlorate Nonahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.