missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, Puriss., ≥98% Al2O3 basis, Honeywell Fluka™

Product Code. 15651010 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
25 kg
6 x 1 kg
Unit Size:
1kg
25kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15651010

missing translation for 'mfr': Honeywell-Fluka 11028500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss.,≥98% Al2O3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Quantity 500 g
Linear Formula Al2O3
UN Number NONH for all modes of transport
Formula Weight 101.96g/mol
Percent Purity ≥98% (Al2O3 basis)
Grade puriss.
Chemical Name or Material Aluminum Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.