missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Powder, Chromatography, BAKER ANALYZED™ Reagent, J.T. Baker™
300.20 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
- Suitable for Chromatographic Use
Specifications
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 101.96 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 4.00kg |
Chemical Name or Material | Aluminum Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'