missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Pellets, 3mm, Honeywell Fluka™

Product Code. 15665450 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15665450 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15665450 Supplier Honeywell-Fluka Supplier No. 4140691KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pellets, 3 mm

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2980°C
Melting Point 2040°C
CAS 1344-28-1
Color Colorless
Quantity 1 kg
Molecular Formula Al2O3
Linear Formula Al2O3
MDL Number MFCD00003424
UN Number NONH for all modes of transport
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
Solubility Information Insoluble in water
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
IUPAC Name dialuminium(3+) trioxidandiide
Molecular Weight (g/mol) 101.96
PubChem CID 9989226
Formula Weight 101.96g/mol
Odor Odorless
Packaging Poly Bottle
Appearance Pellets
Density 3.96g/cm3
Chemical Name or Material Aluminum oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.