missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, neutral, for TLC, Thermo Scientific™

Product Code. 15492597
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15492597 100 g 100g
15402607 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15492597 Supplier Alfa Aesar™ Supplier No. 046056.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For TLC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Al2O3
Merck Index 14,356
Solubility Information Slightly soluble in alkaline solutions. Insoluble in water.
Formula Weight 101.96
Odor Odorless
Refractive Index 1.765
Chemical Name or Material Aluminum oxide, Neutral
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.