missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, neutral, HPLC Flash Grade

Product Code. 15452597
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452597 1 kg 1kg
15442597 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452597 Supplier Thermo Scientific Alfa Aesar Supplier No. 046025.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is an inorganic compound used as a grinding or blending agent. In addition, Aluminum oxide can be essential in protein extraction as a component in the preparation of tissues. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 1 kg
Linear Formula Al2O3
Merck Index 14,356
Solubility Information Insoluble in water.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade HPLC Flash
Odor Odorless
Refractive Index 1.765
Chemical Name or Material Aluminum oxide, Neutral
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.