missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, gamma-phase, catalyst support, high surface area, bimodal

Product Code. 11350488
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350488 1 kg 1kg
11370488 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350488 Supplier Thermo Scientific Alfa Aesar Supplier No. 043832.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Physical Form Pellets
Quantity 1 kg
Solubility Information Insoluble in water. Slightly soluble in acid and alkali.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Packaging Plastic bottle
Specific Gravity 3.965
Density 3.965
Chemical Name or Material Aluminum oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.