missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, fused, insulating powder, 99.7+%

Product Code. 11383128
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383128 1 kg 1kg
11393128 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383128 Supplier Thermo Scientific Alfa Aesar Supplier No. 033199.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in Dry Filling For thermal Insulation of Electrical Furnaces | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Used in dry filling for thermal insulation of electrical furnaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: approx. 180 micron
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Quantity 1 kg
Assay Percent Range >99.7%
IUPAC Name dialuminum;oxygen(2-)
Odor Odorless
Appearance White powder
Vapor Pressure 1.33 hPa (1mmHg) at 2158°C
Chemical Name or Material Aluminum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.