missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Aluminum oxide, Drysphere™ desiccant, without indicator

Brand:  Alfa Aesar™ 046172.22

Code : NEW

Additional Details : CAS Number : 1344-28-1 Weight : 0.10000kg

Quantity 100g

Product Code. 15407165

  • £14.10 / 100g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

Aluminum oxide, Drysphere™ desiccant
1344-28-1
Al2O3
101.96
14,356
6.35mm
2980°C
2045°C
Without indicator
Spheres
Al2O3
MFCD00003424
Odorless
1.765
White
100g
Videos
SDS