missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%), Thermo Scientific Chemicals

Product Code. 11333138
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333138 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11333138 Supplier Thermo Scientific Alfa Aesar Supplier No. 033201.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 60 micron

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Agglomerates
Quantity 1 kg
Assay Percent Range Al2O3 95% (SiO2 ∽5%)
IUPAC Name dialuminium(3+) trioxidandiide
Odor Odorless
Appearance White powder
Vapor Pressure 1.33 hPa (1mmHg) at 2158°C
Chemical Name or Material Aluminum oxide, Contains 0.5% organic binder to increase dry strength.
What are the recommended guidelines for use of the Aluminum oxide, cement, Al2O3 95% (SiO2 ∽5%) (Cat. No. 033201.A1)?

We recommend the following guidelines for use of Aluminum oxide, cement, Al2O3 95% (SiO2 ~5%) (Cat. No. 033201.A1):
Mixing: Add a minimum amount of water until the cement has reach your desired consistency.
Hardening: This product it is not cold-hardening. It contains an organic binder to increase dry strength. The organic binder is completely burned off during the heating process. Above 1200 degrees C, progressive hardening of the cement will occur as a result of sintering.
Curing: The curing time can vary depending on several factors, including: the amount of water used to form the slurry, the temperature used during hardening, and the degree of sintering desired (if heating to temperatures over 1200 degrees C).

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.