missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, alpha-phase, gamma-phase, 99.99% (metals basis)

Product Code. 11392498
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392498 500 g 500g
11382498 25 g 25g
11372498 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11392498 Supplier Thermo Scientific Alfa Aesar Supplier No. 014558.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

It is used in stainless steel coating and in microwave applications. It is used as an adsorbent, desiccating agent, grinding or blending agent, abrasive agent and catalyst, and in the manufacture of dental cements and refractories. Industrial applications include electrical and electronic products, metal Products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Melting Point 2045°C
Physical Form Powder
Quantity 500 g
Merck Index 14,356
Solubility Information Water insoluble. Soluble in hexane,toluene.
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Percent Purity 99.99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Aluminum oxide, alpha-phase, gamma-phase
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.