Learn More
Aluminum oxide, Aerosol Refractory Brushable Paint, Thermo Scientific Chemicals
Description
Provides protective high temperature Al{2}O{3} coating on ceramic, graphite, or metalAluminum oxide, Aerosol Refractory Brushable Paint is used as barrier layer between dissimilar materials to prevent reaction and interactions during high temperature. It also acts as a solvent-based paint. It also provides protective high temperature alumina coating on ceramic, graphite and metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Paste |
Quantity | 250 g |
UN Number | UN1263 |
Flash Point | −4°C (25°F) |
Solubility Information | Slightly soluble in water. |
IUPAC Name | dialuminium(3+) trioxidandiide |
Chemical Name or Material | Aluminum oxide, Aerosol Refractory Brushable Paint |
Safety and Handling
- Aluminum oxide, Aerosol Refractory Brushable Paint
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Ethyl alcohol, Acetone, Hectorite
For Manufacturing and Laboratory Use. Not for beverage use