missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Activated, basic, Brockmann I, Honeywell Fluka™

Product Code. 15661710 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
20 kg
Unit Size:
100g
1kg
20kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15661710 1 kg 1kg
15651710 100 g 100g
15681710 5 kg 5kg
15671710 20 kg 20kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15661710 Supplier Honeywell-Fluka Supplier No. 1994431KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activated, basic, Brockmann I

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

pH Basic
Quantity 1 kg
Linear Formula Al2O3
UN Number NONH for all modes of transport
Formula Weight 101.96g/mol
Chemical Name or Material Aluminum Oxide, Activated Brockmann I
Product Identifier
  • Aluminum oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.