missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, NanoDur™ AL-0405, 99.5%

Product Code. 11450920
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450920 100 g 100g
11460920 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11450920 Supplier Thermo Scientific Alfa Aesar Supplier No. 044931.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

S. A. 32-40m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

NanoDur™ aluminum oxide is designed for coatings where transparency is of utmost importance. NanoDur™ aluminum oxide is especially suited for incorporation into coating systems, greatly enhancing abrasion resistance with minimal effects on cla..Alumium oxide is widely used in a variety of plastics, rubber, ceramics and refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire and yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamps as far-infrared emission and thermal insulation materials. It plays an important role for the hydrogenation of aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5%
Solubility Information Slightly soluble in strong acid and alkaline solution. Insoluble in water.
Formula Weight 101.96
Odor Odorless
Refractive Index 1.768
Appearance White powder
Density 3.965g/cm3
Chemical Name or Material Aluminum oxide, Sold in collaboration with Nanophase Technologies Corporation
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.