missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Aluminum Oxide 90 Acidic

Brand:  Macherey-Nagel™ 815030.1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10105001

  • £55.25 / 1kg
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

50 to 200 μm
4 ± 0.3
∼130 m2/g
Activity
1 kg
Aluminum Oxide 90 Acidic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers