missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxalate hydrate, 99%, Thermo Scientific Chemicals

Catalog Number 11359648
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
This item is not returnable. View return policy
814-87-9
C6Al2O12
318.02
MFCD00050791
ZCLVNIZJEKLGFA-UHFFFAOYSA-H
aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate
93561
dialuminum;oxalate
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
This item is not returnable. View return policy
814-87-9
C6Al2O12
318.02
MFCD00050791
ZCLVNIZJEKLGFA-UHFFFAOYSA-H
aluminum oxalate, dialuminium trioxalate, unii-dq79q766vn, aluminum, tris ethanedioato 2-di, dialuminum oxalate, aluminum oxalate hydrate, dialuminium 3+ trioxalate ion, dialuminium 3+ trioxalate, aluminum oxalate hydrate 10g, dialuminium 3+ ion trioxalate
93561
dialuminum;oxalate
[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Aluminum oxalate is used as a mordant for printing textiles and dyeing cotton. It finds use as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Linear Formula Al2(C2O4)3·xH2O
Quantity 10 g
Merck Index 14,355
Solubility Information Insoluble in water.
Formula Weight 318.02 (Anhydrous)
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material Aluminum oxalate hydrate
compliance-icons
Product Identifier
  • Aluminum oxalate hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'