missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate nonahydrate, ACS, 98.0-102.0%

Product Code. 11343368
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 x 1 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343368 5 x 1 kg 5kg
11333368 50 g 50g
11323368 250 g 250g
11313368 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11343368 Supplier Thermo Scientific Alfa Aesar Supplier No. 036291.D9
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00003419
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73.5°C (135° (decomposition))
Color White
Physical Form Crystalline
Quantity 5 x 1 kg
Assay Percent Range 98 -102%
Solubility Information Very soluble in water and alcohol. Slightly soluble in acetone. Insoluble in ethyl acetate and pyridine.
Formula Weight 375.13 (212.99 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Aluminum nitrate nonahydrate, nonahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.