missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate nonahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Product Code. 11313368
Click to view available options
Quantity:
50 g
250 g
1 kg
5 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00003419
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73.5°C (135° (decomposition))
Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 98 -102%
Solubility Information Very soluble in water and alcohol. Slightly soluble in acetone. Insoluble in ethyl acetate and pyridine.
Formula Weight 375.13 (212.99 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Aluminum nitrate nonahydrate, nonahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'