missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate nonahydrate, 98%

Product Code. 11381739
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
25g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381739 250 g 250g
11351739 25 g 25g
11361739 1 kg 1kg
11371739 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11381739 Supplier Thermo Scientific Alfa Aesar Supplier No. 012360.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol

It finds use in tanning leather, antiperspirant, corrosion protection, extraction of uranium, nitrating agent. It is an effective dopant for producing highly conductive thin film by utilizing the sol gel method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00003419
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73.5°C (135° (decomposition))
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 98%
Solubility Information Soluble in water,ethanol,glycerol and methanol.
Formula Weight 375.13 (212.99 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Aluminum nitrate nonahydrate, nonahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum nitrate nonahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aluminium nitrate nonahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.