missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg)

Product Code. 11340809
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340809 25 g 25g
11330809 5 g 5g
11320809 100 g 100g
11350809 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11340809 Supplier Thermo Scientific Alfa Aesar Supplier No. 010626.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25838-59-9 | AlN3O9 | 212.99 g/mol

Aluminum nitrate hydrate is used to produce alumina for the preparation of insulating papers, and in the extraction of actinide elements. It is also used in the preparation of rare earth aluminates, in the heating elements of cathode ray tubes, as a precursor for sol-gel synthesis of alumina, and on transformer core laminates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25838-59-9
Molecular Formula AlN3O9
Molecular Weight (g/mol) 212.99
MDL Number MFCD00003419
InChI Key JLDSOYXADOWAKB-UHFFFAOYSA-N
Synonym aluminum nitrate hydrate, al.3no3.h2o, aluminum nitrate hydrate, puratronic, aluminum 3+ hydrate trinitrate
PubChem CID 22078336
SMILES [Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 73.5°C
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.999% (Metals basis excluding Hg)
Solubility Information Soluble in water,methanol,ethanol,and ethylene glycol.
IUPAC Name aluminum;trinitrate;hydrate
Formula Weight 212.99 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Aluminum nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.