missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nickel isopropoxide, 99.9% (metals basis), Thermo Scientific™

Product Code. 11314498
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70504-57-3
MDL Number MFCD00143593

Specifications

Quantity 2 g
UN Number UN3181
Sensitivity Moisture sensitive
Formula Weight 584.92
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Aluminum nickel isopropoxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aluminum nickel isopropoxide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'