missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nickel isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol, Thermo Scientific™

Product Code. 11368028
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368028 100 mL 100mL
11378028 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368028 Supplier Thermo Scientific Alfa Aesar Supplier No. 036590.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70504-57-3
MDL Number MFCD00143593
Synonym Nickel aluminum isopropoxide; Aluminum nickel isopropoxide

Specifications

Color Green
Boiling Point 120°C (0.6 mmHg)
Quantity 100 mL
UN Number UN1219
Sensitivity Moisture Sensitive
Formula Weight 584.92
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Aluminum nickel isopropoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.