missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum metaphosphate, Al{2}O{3} 19.4%, P{2}O{5} 78.9%, Thermo Scientific Chemicals
14.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 x 1 kg
Description
Aluminum metaphosphate is used as a constituent of glazes, enamels, glasses and as a high-temperature insulating cement. It is also used as molecular sieves.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Insoluble in water. |
Formula Weight | 263.9 |
Concentration or Composition (by Analyte or Components) | Al2O3 19.4%, P2O5 78.9% |
Density | 2.779 g/mL |
Chemical Name or Material | Aluminum metaphosphate |
Safety and Handling
Product Identifier
- Aluminum metaphosphate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- EUH014-Reacts violently with water.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'