missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Aluminum Lids for Aluminum Basin

Nickel Electro™ Aluminun Lid for aluminum Basin is crevice free and one piece construction.

Brand:  Nickel Electro™ 644

Code : 03

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12717356

  • £46.95 / Pack of 5
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Nickel Electro™ Aluminun Lid for aluminum Basin is crevice free and one piece construction.

  • Aluminum
  • Seamless
  • One piece construction
  • Crevice free
  • For use with aluminum basin
Specifications

Specifications

Aluminum
Lid
5 mm
75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers