missing translation for 'onlineSavingsMsg'
Learn More

Aluminum lactate, 90-95%, contains ^=5% water

Product Code. 11303508
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11303508

Brand: Thermo Scientific Alfa Aesar 039122.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18917-91-4 | C9H15AlO9 | 294.19 g/mol

It is used as a skin moisturizer lotion to treat dry, scaly, itchy skin, reagent in biological activity studies of aluminum and in sol-gel processing. Also used in fire foam. Aluminum L-Lactate has been used to inject aluminum into rabbits. Aluminum L-Lactate has been shown to decrease human enamel dissolution in a pH 4 acetate buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18917-91-4
Molecular Formula C9H15AlO9
Molecular Weight (g/mol) 294.19
MDL Number MFCD20529175
InChI Key VXYADVIJALMOEQ-UHFFFAOYNA-K
IUPAC Name bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate
SMILES CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O

Specifications

Physical Form Powder
Quantity 500 g
Merck Index 14,348
Solubility Information Very soluble in cold water
Formula Weight 294.2
Percent Purity 90 to 95%
Concentration or Composition (by Analyte or Components) Contains ≈5% water
Odor Odorless
Chemical Name or Material Aluminum lactate
compliance-icons
Product Identifier
  • Aluminum lactate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Aluminum lactate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.