missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Isopropoxide, TRC

Product Code. 30316055
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316055 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316055 Supplier Toronto Research Chemicals Supplier No. TRCA5759301KG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum Isopropoxide
CAS 555-31-7
Molecular Formula C9H24AlO3
Quantity 1 kg
Synonym 2-Propanol Aluminum Salt, Isopropyl Alcohol Aluminum Salt, AIPD, Aliso, Aluminum 2-Propoxide, Aluminum Isopropanolate, Aluminum Isopropioate, Aluminum Isopropylate, Aluminum sec-Propanolate, Aluminum Triisopropoxide, Aluminum Triisopropylate, Aluminum Tris(isopropoxide), Aluminum Tris(isopropylate), Aluminum Tris(sec-propoxide), Aluminum(3+) Isopropoxide, Isopropanol Aluminum Salt, Manalox 130, PADM, SYM-AL 04, Triisopropoxyaluminum, Triisopropyl Aluminate, Triisopropyloxyaluminum, Tris(isopropanolato)aluminum, Tris(isopropoxy)aluminum
InChI Formula InChI=1S/3C3H7O.Al/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3
SMILES CC(C)O[Al](OC(C)C)OC(C)C
IUPAC Name tri(propan-2-yloxy)alumane
Molecular Weight (g/mol) 207.27
Formula Weight 204.1306
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.