missing translation for 'onlineSavingsMsg'
Learn More

Aluminum isopropoxide, 98+%

Product Code. 11301539
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11301539

Brand: Thermo Scientific Alfa Aesar 014007.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for the reduction of carbonyl compounds | CAS: 555-31-7 | C9H21AlO3 | 204.246 g/mol

Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates. Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reagent for the reduction of carbonyl compounds
  • Used in alcoholysis and ester exchange reactions
  • Also used in synthesis of higher alkoxides, chelates, and acrylates
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-31-7
Molecular Formula C9H21AlO3
Molecular Weight (g/mol) 204.246
MDL Number MFCD00008870
InChI Key SMZOGRDCAXLAAR-UHFFFAOYSA-N
Synonym aluminum isopropoxide, aluminum isopropylate, aluminum triisopropoxide, triisopropoxyaluminum, aliso, aluminum triisopropanolate, 2-propanol, aluminum salt, triisopropyloxyaluminum, aluminum triisopropylate
PubChem CID 11143
IUPAC Name aluminum;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Al+3]

Specifications

Melting Point 131°C to 135°C
Density 1.035 g/mL
Boiling Point 135°C (10 mmHg)
Assay Percent Range 98+%
Refractive Index 1.0346
Quantity 5 kg
UN Number UN3181
Sensitivity Moisture sensitive
Merck Index 14,347
Solubility Information Soluble in alcohol,benzene,and toluene; Decomposes in water;Soluble in ethanol,isopropanol,benzene,toluene,chloroform,carbon tetrachloride and petroleum hydrocarbons.
Formula Weight 204.25
Percent Purity ≥98%
Physical Form Granules
Chemical Name or Material Aluminum isopropoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.