missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum: ISO Safety Sign - Use handrail

Sign compliant to ISO 7010.

Brand:  Brady™ 819621

Additional Details : Weight : 0.09300kg

 View more versions of this product

Product Code. 15694507

  • £15.05 / Each
Estimated Shipment: 12-03-2024
to see stock.
Add to basket

Description

Description

  • Aluminum
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 140°C
Global
Aluminum
200 mm
1.016 mm
Round
White on Blue
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers