missing translation for 'onlineSavingsMsg'
Learn More

Aluminum iodide, 99.99+% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4781766
Click to view available options
Quantity:
5 g
25 g
7784-23-8
AlI3
407.695
MFCD00003427
CECABOMBVQNBEC-UHFFFAOYSA-K
aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
82222
triiodoalumane
[Al](I)(I)I
7784-23-8
AlI3
407.695
MFCD00003427
CECABOMBVQNBEC-UHFFFAOYSA-K
aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
82222
triiodoalumane
[Al](I)(I)I

Moisture and Light Sensitive

Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 360°C
Melting Point 191°C
Color Brown-Purple to White
Physical Form Lump
Quantity 5 g
Assay Percent Range >99.99% (Metals basis)
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 407.69
Odor Pungent
Sensitivity Light and moisture sensitive
Density 3.98
Chemical Name or Material Aluminum iodide
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum iodide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Aluminum iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'