missing translation for 'onlineSavingsMsg'
Learn More

Aluminum iodide, 99.99+% (metals basis)

Product Code. 11399399
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399399 25 g 25g
11389399 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399399 Supplier Thermo Scientific Alfa Aesar Supplier No. 089185.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Moisture and Light Sensitive | CAS: 7784-23-8 | AlI3 | 407.695 g/mol

Aluminum iodide is employed as a catalyst to break certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides It can also be used for the manufacture of iodinated organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7784-23-8
Molecular Formula AlI3
Molecular Weight (g/mol) 407.695
MDL Number MFCD00003427
InChI Key CECABOMBVQNBEC-UHFFFAOYSA-K
Synonym aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
PubChem CID 82222
IUPAC Name triiodoalumane
SMILES [Al](I)(I)I

Spécification

Boiling Point 360°C
Melting Point 191°C
Color Brown-Purple to White
Physical Form Lump
Quantity 25 g
Assay Percent Range >99.99% (Metals basis)
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 407.69
Odor Pungent
Sensitivity Light and moisture sensitive
Density 3.98 g/cm3
Chemical Name or Material Aluminum iodide
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.