missing translation for 'onlineSavingsMsg'
Learn More

Aluminum iodide, ultra dry, 99.999% (metals basis)

Product Code. 15432887
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15432887 Supplier Thermo Scientific Alfa Aesar Supplier No. 047170.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7784-23-8 | AlI3 | 407.695 g/mol

It is employed as a reagent for the scission of certain kinds of C-O and N-O bonds. It cleaves aryl ethers and deoxygenates epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Spezifikation

Boiling Point 360°C
Melting Point 191°C
CAS 7784-23-8
Color Brown-Purple to White
Physical Form Powder
Quantity 5 g
Odor Pungent
Packaging Ampule
Sensitivity Light and moisture sensitive
Density 3.98 g/mL
Molecular Formula AlI3
MDL Number MFCD00003427
UN Number UN3260
Merck Index 14,346
Synonym aluminum iodide, aluminum triiodide, aluminum iii iodide, aluminum iodide anhydrous, aluminum iodide, technical grade, aluminum triiodide, aluminum iodide metals basis, aluminum iodide, anhydrous, powder trace metals basis
Solubility Information Reacts with water.
InChI Key CECABOMBVQNBEC-UHFFFAOYSA-K
SMILES [Al](I)(I)I
IUPAC Name triiodoalumane
Molecular Weight (g/mol) 407.695
PubChem CID 82222
Formula Weight 407.69
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Aluminum iodide, Ultra dry
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.