missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Hydroxide, Reagent grade, Honeywell™

Product Code. 15612420 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
25g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15612420 25 g Poly Bottle 25g
15632420 500 g Poly Bottle 500g
15622420 2 kg Poly Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15612420 Supplier Honeywell Research Chemicals Supplier No. 23918625G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21645-51-2
Molecular Formula AlH3O3
Molecular Weight (g/mol) 78.00
MDL Number MFCD00003420
InChI Key WNROFYMDJYEPJX-UHFFFAOYSA-K
IUPAC Name aluminium(3+) trihydroxide
SMILES [OH-].[OH-].[OH-].[Al+3]

Specifications

Melting Point 300°C
Color White
Quantity 25 g
Linear Formula Al(OH)3
UN Number NONH for all modes of transport
Formula Weight 78g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.42g/cm3
Chemical Name or Material Aluminum Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.