Learn More
Wesbart™ Aluminum Horizontal Freezer Rack
Wesbart™ Aluminum Horizontal Freezer Racks are front loading with trays for quick access to cryoboxes and other stored items.
Brand:  Wesbart™ BH115/2NMT
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 16224941
  • £106.00 / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Aluminum front access storage rack for tray units (ordered separately, Cat. No. 17311849) for upright freezers
  • For standard cryobox or other freezer storage
  • Only one tray has to be removed from the rack at a time for sample access, minimizing temperature impact on remaining items
  • Range of sizes and capacities available
Specifications
Specifications
Freezer Storage Rack
Aluminum
Upright Freezers
Boxes (10/Rack)
2 Trays/Rack
685 x 140 x 287 mm