missing translation for 'onlineSavingsMsg'
Learn More

Aluminum hexafluoro-2,4-pentanedionate, 98+%, Thermo Scientific™

Product Code. 11309498
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309498 2 g 2g
11399488 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309498 Supplier Thermo Scientific Alfa Aesar Supplier No. 043328.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

In MOCVD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15306-18-0
Molecular Formula C15H3AlF18O6
Molecular Weight (g/mol) 648.14
MDL Number MFCD00050051
InChI Key SINRSQHVMMWHSI-UHFFFAOYSA-N
Synonym aluminum hexafluoroacetylacetonate, aluminum hexafluoropentanedionate, aluminum hexafluoroacetylacetonate, 3z-4-bis 2z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-yl oxy alumanyl oxy-1,1,1,5,5,5-hexafluoropent-3-en-2-one
IUPAC Name aluminium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
SMILES [Al+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F

Specifications

Boiling Point 170°C (decomposition)
Melting Point 70°C to 73°C
Physical Form Crystalline
Quantity 2 g
Formula Weight 648.17
Percent Purity ≥98%
Sensitivity Moisture sensitive
Chemical Name or Material Aluminum hexafluoro-2,4-pentanedionate
compliance-icons
Product Identifier
  • Aluminum hexafluoro-2,4-pentanedionate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.