missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ 96-well Microtitration Plates, Aluminum

Product Code. 10251152
Click to view available options
Total Volume (Metric) Well:
1.2 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10251152

Brand: Hirschmann™ 9242196

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Microplates, deep well, with glass inserts in common plates formats. Glass providing excellent chemical inertness, no leaching of plastics.

  • Available with fixed or loosely inserted glass inserts
  • Highly pure
  • Chemically inert

Specifications

Format Standard
Lid With Lid
Array 8 x 12
For Use With (Equipment) Loose Glaskavit?ten
Material Glass
No. of Wells 96
Total Volume (Metric) Well 1.2 mL
For Use With (Application) Loose Glaskavit?ten
Includes Transparent cover and 96 Cavities
No. per Pack 1 Each
Quantity Each
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.