missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, alloy 6061, 2.0mm (0.08in) thick

Product Code. 15402187
Click to view available options
Quantity:
300 x 300 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15402187

Brand: Thermo Scientific Alfa Aesar 045171.HB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

Aluminum foil, alloy 6061 is commonly used for structural components, frames, brackets, jigs, fixtures, base plates, machine parts, hydraulic valve bodies, valves parts, fuse parts, gears, worm gears, rectifier parts, fasteners, truck and marine components, marine fittings and hardware, electrical fittings and connectors, hinge pins, magneto parts, appliance fittings, camera lens and mounts 6061 is used for heavy duty structures requiring good strength-to-weight ratio with good corrosion resistance. 6061 is easily cold worked and formed in the annealed condition. Cutting, stamping, bending, deep drawing, drilling and tapping. are all readily accomplished using standard methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum foil, Alloy 6061
Name Note 2.0mm (0.08 in.) thick
Quantity 300 x 300 mm
Odor Odorless
Form Foil
MDL Number MFCD00134029
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.