missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, alloy 6061, 1.27mm (0.05in) thick

Product Code. 15422217
Change view
Click to view available options
Quantity:
300 x 300 mm
Unit Size:
Each
Product Code. Quantity unitSize
15422217 300 x 300 mm Each
1 options
This item is not returnable. View return policy

Product Code. 15422217

Brand: Thermo Scientific Alfa Aesar 045226.HB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Silver gray foil
Quantity 300 x 300 mm
Odor Odorless
MDL Number MFCD00134029
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.