Learn More
Aluminum foil, alloy 6061, 1.0mm (0.04in) thick
Description
Aluminum foil, alloy 6061 is commonly used for structural components, frames, brackets, jigs, fixtures, base plates, machine parts, hydraulic valve bodies, valves parts, fuse parts, gears, worm gears, rectifier parts, fasteners, truck and marine components, marine fittings and hardware, electrical fittings and connectors, hinge pins, magneto parts, appliance fittings, camera lens mounts, etc. 6061 is used for heavy duty structures requiring good strength-to-weight ratio with good corrosion resistance. 6061 is easily cold worked and formed in the annealed condition. Cutting, stamping, bending, deep drawing, drilling, tapping, etc. are all readily accomplished using standard methods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Aluminum foil, Alloy 6061 |
| Name Note | 1.0mm (0.04 in.) thick |
| Quantity | 300 x 300 mm |
| Odor | Odorless |
| Form | Foil |
| MDL Number | MFCD00134029 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.