Learn More
Alfa Aesar™ Aluminum foil, 1.5mm (0.06 in.) thick, Puratronic™, 99.998% (metals basis)
Brand: Alfa Aesar™ 044492.HP
Code : NEW
Additional Details : CAS Number : 7429-90-5 Weight : 0.01000kg
Quantity | 100x500mm |
---|
Chemical Identifiers
Aluminum foil | |
1.5mm (0.06 in.) thick, Temper: as rolled | |
MFCD00134029 |
7429-90-5 | |
Puratronic™ |
Specifications
Aluminum foil | |
100x500mm | |
7429-90-5 | |
Foil | |
(metals basis) | |
Odorless |
≈10.125g/50x50mm | |
1.5mm (0.06 in.) thick, Temper: as rolled | |
Puratronic™ | |
99.998% | |
MFCD00134029 |