missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Fluoride Trihydrate, 97%, Honeywell Fluka™

Product Code. 15632330 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15632330 100 g Poly Bottle 100g
15642330 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15632330 Supplier Honeywell-Fluka Supplier No. 236098100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3H6O3
Molecular Weight (g/mol) 138.022
MDL Number MFCD00149141
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]

Specifications

Quantity 100 g
Linear Formula AlF3·3H2O
UN Number NONH for all modes of transport
Formula Weight 138.02g/mol
Percent Purity 97%
Packaging Poly Bottle
Density 2.88g/mL at 25°C
Chemical Name or Material Aluminum Fluoride Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.