missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum fluoride, anhydrous, 99.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Aluminum fluoride (AlF3) is an inorganic compound used primarily in the production of aluminum. It is also used to inhibit fermentation. It is a sputtering target for preparation of low index films. In ceramics, as flux in metallurgy, in aluminum manufacture, as inhibitor of fermentation, as catalyst in organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1291°C (sublimation) |
Physical Form | Sintered Lump |
Quantity | 100 g |
Merck Index | 14,339 |
Solubility Information | Sparingly soluble in acids and alkalies. Insoluble in Acetone. |
Formula Weight | 83.98 |
Percent Purity | 99.5% |
Sensitivity | Hygroscopic |
Density | 2.882 g/mL |
Chemical Name or Material | Aluminum fluoride, Anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'