missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Aluminum Fixed Angle Rotors

Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.

Brand:  Eppendorf™ 5427716009

Code : 99

Additional Details : Weight : 4.95000kg

 View more versions of this product

Product Code. 10368522

  • £1315.80 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Lid (see details for type)

Description

Description

Aerosol-tight rotors (05-401-503, 05-400-023 and 05-401-505) feature a distinct red rotor lid screw and red ring inside the rotor for easy identification. Tube adapters are available separately.

Standard 30-place Rotors

  • Capacity: 30 × 1.5/2.0mL Molecular-grade Max. Speed/RCF: 14,000rpm/20,8170xG
  • Available with aerosol-tight lid for use with hazardous samples (05-401-503), or with plastic lid for basic applications (05-400-019)
  • PTFE-coated for high chemical resistance and easy cleaning
24-place High-speed Fixed-angle Rotor
  • PTFE-coated for high chemical resistance and easy cleaning
  • Capacity: 24 × 1.5/2.0mL tubes
  • Max. Speed/RCF: 17,500rpm/30,1300xG
  • Includes aerosol-tight lid for hazardous sample work
  • Check tube stability before use; Eppendorf 1.5mL Flex-Tube™ or Safe-Lock™ Tubes recommended
2-place Swinging Bucket Microplate Rotor
  • Safely spins two standard microplates
  • Max. Speed/RCF: 4680rpm/22,040xG
  • Loading height of 29mm is suitable for all types of MTP or PCR plates
  • Windshielded design and drop-on lid ensure virtually silent centrifugation

15mL/50mL Conical Tube Rotor
  • Capacity: 6 × 15mL or 6 × 50mL conical tubes
  • Max. Speed/RCF: 7830rpm/7,197xG
  • Adapters for 15 and 50mL conical tubes included
  • 50mL adapter also accommodates 50mL skirted conical tubes

24-place Spin Column Kit Rotor
  • Capacity: 24 × spin columns or 24 × 1.5/2.0mL tubes
  • Max. Speed/RCF: 13,200rpm/19,090xG
  • Unique high rim design supports open tube lids during centrifugation; lids will not tear off
  • Aerosol-tight lid for hazardous samples

PCR Strip Rotor
  • Capacity: 8 × 8 PCR tube strips or 64 × 0.2mL PCR tubes
  • Max. Speed/RCF: 11,800rpm/13,5480xG
  • Includes removable adapters that can be used as a tube rack for pipetting

Cryotube Rotor
  • Ideal for tissue culture labs that need to spin down samples before cryo conservation
  • Accepts up to 18 cryo vials
  • Max. Speed/RCF: 8900rpm/83,200xG
  • Includes adapters to accommodate tubes with 16.9 or 13.4mm diameter

 

Certifications

Aerosol-tight rotors are tested and certified by the Health Protection Agency (Porton Down, UK)

Specifications

Specifications

PTFE Coated
6 x 15/50 mL
7,830 rpm
5430 Microcentrifuge
Screw-on lid
Rotor
7,197 x g
9.5 cm
Fixed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers