missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ethoxide

Product Code. 11334048
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11334048

Brand: Thermo Scientific Alfa Aesar 041336.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 555-75-9 | C6H15AlO3-3 | 162.165 g/mol

Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-75-9
Molecular Formula C6H15AlO3-3
Molecular Weight (g/mol) 162.165
MDL Number MFCD00009658
InChI Key JBRFPFQROJYWFN-UHFFFAOYSA-N
Synonym acmc-209lnt
PubChem CID 70700059
IUPAC Name aluminum;ethanolate
SMILES CC[O-].CC[O-].CC[O-].[Al]

Specifications

Melting Point 154°C to 159°C
Color White
Density 1.142 g/mL
Boiling Point 320°C
Linear Formula Al(OC2H5)3
Quantity 250 g
UN Number UN2925
Sensitivity Moisture sensitive
Merck Index 14,338
Solubility Information Soluble in hot xylene,cholorobenzene. Reacts with water.
Formula Weight 162.16
Physical Form Chunks and/or Powder
Chemical Name or Material Aluminum ethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.