missing translation for 'onlineSavingsMsg'
Learn More

Aluminum dihydrogen phosphate, 50% w/w aqueous solution, Thermo Scientific Chemicals

Product Code. 11300098
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11300098

Brand: Thermo Scientific Alfa Aesar 042858.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aluminum dihydrogen phosphate is used as intermediates and solid separation agents. It is also used in petroleum, chemical industry, shipbuilding, space technology, electric industry, high-temperature kilns, heat treatment resistance furnaces and electric insulation. Further, it is used as inorganic coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13530-50-2
Molecular Formula AlH6O12P3
Molecular Weight (g/mol) 317.94
MDL Number MFCD00014152
InChI Key RGPUVZXXZFNFBF-UHFFFAOYSA-K
Synonym aluminum dihydrogen phosphate, aluminum phosphate monobasic, prim-aluminum phosphate, aluminum tris dihydrogen phosphate, phosphoric acid, aluminum salt 3:1, aluminum hydrogen phosphate, aluminumdihydrogen phosphate, aluminum phosphate,monobasic, aluminum trisdihydrogenphosphate, aluminum dihydrogen orthophosphate
PubChem CID 9818426
IUPAC Name aluminum;dihydrogen phosphate
SMILES [Al+3].OP(O)([O-])=O.OP(O)([O-])=O.OP(O)([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 5 kg
Assay Percent Range 50% w/w aqueous solution
Solubility Information Soluble in water and ethanol. Insoluble in acetone.
Formula Weight 317.94
Appearance Colorless liquid
Density 1.5 g/cm3
Chemical Name or Material Aluminum dihydrogen phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.